Learn More
ACROBiosystems Biotinylated Human Nectin-2 / CD112 Protein, Fc,Avitag™
Catalog No. 502055316
Encompass_Preferred
Recombinant Protein;25UG;Biotinylated Human Nectin-2, Fc,Avitag (CD2-H82F8) is expressed from human 293 cells (HEK293). It contains AA Gln 32 - Leu 360 (Accession # Q92692-2).;This protein carries a human IgG1 Fc tag at the C-terminus, followed by a Avi tag (Avitag™). The protein has a calculated MW of 64.0 kDa. As a result of glycosylation, the protein migrates as 66 kDa under reducing (R) condition, and 130 kDa under non-reducing (NR) condition (SDS-PAGE).;Immobilized Human PVRIG, Mouse IgG2a Fc Tag, low endotoxin (Cat. No. PVG-H5253) at 2 μg/mL (100 μL/well) can bind Biotinylated Human Nectin-2, Fc,Avitag (Cat. No. CD2-H82F8) with a linear range of 0.2-13 ng/mL (QC tested).;Immobilized Human TIGIT, Fc Tag (Cat. No. TIT-H5254) at 10 μg/mL (100 μL/well) can bind Biotinylated Human Nectin-2, Fc,Avitag (Cat. No. CD2-H82F8) with a linear range of 0.313-10 μg/mL (Routinely tested).
Catalog No. 50-205-5316 Supplier ACROBiosystems Supplier No. CD2H82F825UG
Recombinant Protein;25UG;Biotinylated Human Nectin-2, Fc,Avitag (CD2-H82F8) is expressed from human 293 cells (HEK293). It contains AA Gln 32 - Leu 360 (Accession # Q92692-2).;This protein carries a human IgG1 Fc tag at the C-terminus, followed by a Avi tag (Avitag™). The protein has a calculated MW of 64.0 kDa. As a result of glycosylation, the protein migrates as 66 kDa under reducing (R) condition, and 130 kDa under non-reducing (NR) condition (SDS-PAGE).;Immobilized Human PVRIG, Mouse IgG2a Fc Tag, low endotoxin (Cat. No. PVG-H5253) at 2 μg/mL (100 μL/well) can bind Biotinylated Human Nectin-2, Fc,Avitag (Cat. No. CD2-H82F8) with a linear range of 0.2-13 ng/mL (QC tested).;Immobilized Human TIGIT, Fc Tag (Cat. No. TIT-H5254) at 10 μg/mL (100 μL/well) can bind Biotinylated Human Nectin-2, Fc,Avitag (Cat. No. CD2-H82F8) with a linear range of 0.313-10 μg/mL (Routinely tested).