Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ACROBiosystems Biotinylated Human Nectin-4 (154-243) Protein, His, Avi tag (NE4-H82Ec)

Catalog No. NC3350319
Encompass_Preferred

Recombinant biotinylated extracellular fragment of human Nectin-4 (Asn 154-Ser 243) expressed in HEK293 cells. The protein includes an N-terminal polyhistidine tag and an Avi tag for site-specific enzymatic biotinylation, and is supplied as a glycosylated protein suitable for binding and detection assays.

  • Expressed in HEK293 cells for mammalian glycosylation.
  • Contains amino acids Asn 154-Ser 243 of human Nectin-4.
  • N-terminal polyhistidine and Avi tag for purification and site-specific biotinylation.
  • Enzymatically biotinylated at the Avi tag for streptavidin-based capture or detection.
  • Glycosylated; apparent molecular weight 15-30 kDa on SDS-PAGE.
  • Purity >90% by SDS-PAGE.
  • Suitable for binding assays, ELISA, SPR, and receptor interaction studies.

Catalog No. NC3350319 Supplier ACROBiosystems Supplier No. NE4H82EC
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Recombinant biotinylated extracellular fragment of human Nectin-4 (Asn 154-Ser 243) expressed in HEK293 cells. The protein includes an N-terminal polyhistidine tag and an Avi tag for site-specific enzymatic biotinylation, and is supplied as a glycosylated protein suitable for binding and detection assays.

  • Expressed in HEK293 cells for mammalian glycosylation.
  • Contains amino acids Asn 154-Ser 243 of human Nectin-4.
  • N-terminal polyhistidine and Avi tag for purification and site-specific biotinylation.
  • Enzymatically biotinylated at the Avi tag for streptavidin-based capture or detection.
  • Glycosylated; apparent molecular weight 15-30 kDa on SDS-PAGE.
  • Purity >90% by SDS-PAGE.
  • Suitable for binding assays, ELISA, SPR, and receptor interaction studies.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.