Learn More
ACROBiosystems ACROBiosystems Biotinylated Human PD-1 / PDCD1 Protein, Mouse IgG2a Fc,Avitag™ (MALS verified)
Catalog No. 502106215
Encompass_Preferred
Recombinant Protein;200UG;Biotinylated Human PD-1, Mouse IgG2a Fc,Avitag (PD1-H82A4) is expressed from human 293 cells (HEK293). It contains AA Leu 25 - Gln 167 (Accession # Q15116-1).;This protein carries a mouse IgG2a Fc tag at the C-terminus, followed by an Avi tag (Avitag™). The protein has a calculated MW of 44.5 kDa. The protein migrates as 60-70 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Human PD-L1, Fc Tag (Cat. No. PD1-H5258) at 2 μg/mL (100 μL/well) can bind Biotinylated Human PD-1, Mouse IgG2a Fc,Avitag (Cat. No. PD1-H82A4) with a linear range of 5-156 ng/mL (QC tested).;Immobilized Nivolumab at 2 μg/mL (100 μL/well) can bind Biotinylated Human PD-1, Mouse IgG2a Fc,Avitag (Cat. No. PD1-H82A4) with a linear range of 0.2-5 ng/mL (Routinely tested).
Catalog No. 50-210-6215 Supplier ACROBiosystems Supplier No. PD1H82A4200UG
Recombinant Protein;200UG;Biotinylated Human PD-1, Mouse IgG2a Fc,Avitag (PD1-H82A4) is expressed from human 293 cells (HEK293). It contains AA Leu 25 - Gln 167 (Accession # Q15116-1).;This protein carries a mouse IgG2a Fc tag at the C-terminus, followed by an Avi tag (Avitag™). The protein has a calculated MW of 44.5 kDa. The protein migrates as 60-70 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Human PD-L1, Fc Tag (Cat. No. PD1-H5258) at 2 μg/mL (100 μL/well) can bind Biotinylated Human PD-1, Mouse IgG2a Fc,Avitag (Cat. No. PD1-H82A4) with a linear range of 5-156 ng/mL (QC tested).;Immobilized Nivolumab at 2 μg/mL (100 μL/well) can bind Biotinylated Human PD-1, Mouse IgG2a Fc,Avitag (Cat. No. PD1-H82A4) with a linear range of 0.2-5 ng/mL (Routinely tested).