Learn More
ACROBiosystems ACROBiosystems Biotinylated Human PD-L1 / B7-H1 Protein, Mouse IgG2a Fc,Avitag™ (MALS verified)
Catalog No. 502106238
Encompass_Preferred
Recombinant Protein;200UG;Biotinylated Human PD-L1, Mouse IgG2a Fc,Avitag (PD1-H82A3) is expressed from human 293 cells (HEK293). It contains AA Phe 19 - Arg 238 (Accession # NP_054862.1).;This protein carries a mouse IgG2a Fc tag at the C-terminus, followed by an Avi tag (Avitag™). The protein has a calculated MW of 53.8 kDa. The protein migrates as 65-80 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Human PD-1, Fc Tag (Cat. No. PD1-H5257) at 2 μg/mL (100 μL/well) can bind Biotinylated Human PD-L1, Mouse IgG2a Fc,Avitag (Cat. No. PD1-H82A3) with a linear range of 5-39 ng/mL (QC tested).
Catalog No. 50-210-6238 Supplier ACROBiosystems Supplier No. PD1H82A3200UG
Recombinant Protein;200UG;Biotinylated Human PD-L1, Mouse IgG2a Fc,Avitag (PD1-H82A3) is expressed from human 293 cells (HEK293). It contains AA Phe 19 - Arg 238 (Accession # NP_054862.1).;This protein carries a mouse IgG2a Fc tag at the C-terminus, followed by an Avi tag (Avitag™). The protein has a calculated MW of 53.8 kDa. The protein migrates as 65-80 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Human PD-1, Fc Tag (Cat. No. PD1-H5257) at 2 μg/mL (100 μL/well) can bind Biotinylated Human PD-L1, Mouse IgG2a Fc,Avitag (Cat. No. PD1-H82A3) with a linear range of 5-39 ng/mL (QC tested).