Learn More
ACROBiosystems Biotinylated Human PD-L2 Protein, Avitag™, His Tag, 25 μg
Catalog No. 502106774
Encompass_Preferred
This biotinylated human PD-L2/B7-DC protein is expressed in human HEK293 cells and carries a C-terminal Avitag™ and polyhistidine tag. As a member of the immunoglobulin superfamily, PD-L2 is essential for T-cell proliferation and cytokine production. This protein is biotinylated enzymatically at the single lysine residue within the Avitag. It is supplied as a lyophilized powder and is validated for use in binding experiments such as biopanning, though it is not recommended for PD1-PDL2 binding applications.
  • Greater than 95% purity as determined by SDS-PAGE
  • Calculated molecular weight of 25.2 kDa
  • Lyophilized from a 0.22 μm filtered PBS solution, pH 7.4, with trehalose protectant
  • Validated by ELISA for activity
  • Stable for 12 months at -20°C to -70°C in lyophilized state
Catalog No. 50-210-6774 Supplier ACROBiosystems Supplier No. PD2H82E825UG
This biotinylated human PD-L2/B7-DC protein is expressed in human HEK293 cells and carries a C-terminal Avitag™ and polyhistidine tag. As a member of the immunoglobulin superfamily, PD-L2 is essential for T-cell proliferation and cytokine production. This protein is biotinylated enzymatically at the single lysine residue within the Avitag. It is supplied as a lyophilized powder and is validated for use in binding experiments such as biopanning, though it is not recommended for PD1-PDL2 binding applications.
  • Greater than 95% purity as determined by SDS-PAGE
  • Calculated molecular weight of 25.2 kDa
  • Lyophilized from a 0.22 μm filtered PBS solution, pH 7.4, with trehalose protectant
  • Validated by ELISA for activity
  • Stable for 12 months at -20°C to -70°C in lyophilized state