Learn More
ACROBiosystems ACROBiosystems Biotinylated Human Prolactin / PRL Protein, Fc,Avitag™
Supplier:  ACROBiosystems PRNH82F7200UG
Encompass_Preferred
Recombinant Protein;200UG;Biotinylated Human Prolactin, Fc,Avitag (PRN-H82F7) is expressed from human 293 cells (HEK293). It contains AA Leu 29 - Cys 227 (Accession # Q5THQ0-1).;This protein carries a human IgG1 Fc tag at the C-terminus, followed by a Avi tag (Avitag™). The protein has a calculated MW of 51.2 kDa. The protein migrates as 57-66 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Human Prolactin R, Fc Tag (Cat. No. PRP-H5251) at 5 μg/mL (100 μL/well) can bind Biotinylated Human Prolactin, Fc,Avitag (Cat. No. PRN-H82F7) with a linear range of 5-156 ng/mL (QC tested).
Catalog No. 50-210-6714