Learn More
ACROBiosystems ACROBiosystems Biotinylated Human PSGL-1 / CD162 Protein, His,Avitag™
Catalog No. 502106518
Encompass_Preferred
Recombinant Protein;200UG;Biotinylated Human PSGL-1, His,Avitag (PS1-H82E4) is expressed from human 293 cells (HEK293). It contains AA Gln 42 - Cys 320 (Accession # Q14242-1).;This protein carries a polyhistidine tag at the C-terminus, followed by an Avi tag. The protein has a calculated MW of 32.4 kDa. The protein migrates as 55-90 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.
Catalog No. 50-210-6518 Supplier ACROBiosystems Supplier No. PS1H82E4200UG
Recombinant Protein;200UG;Biotinylated Human PSGL-1, His,Avitag (PS1-H82E4) is expressed from human 293 cells (HEK293). It contains AA Gln 42 - Cys 320 (Accession # Q14242-1).;This protein carries a polyhistidine tag at the C-terminus, followed by an Avi tag. The protein has a calculated MW of 32.4 kDa. The protein migrates as 55-90 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.