Learn More
ACROBiosystems Biotinylated Human Respiratory Syncytial Virus A (Strain A2) Pre-F0 Protein, His-AviTag, 25 μg
Catalog No. 502357362
Encompass_Preferred
Biotinylated prefusion F0 protein from Human respiratory syncytial virus A (strain A2), engineered with a C-terminal polyhistidine tag and an Avi tag for site-specific enzymatic biotinylation. The protein is expressed in HEK293 cells, MALS-verified, and supplied lyophilized (25 μg) for assay development, binding studies, and antibody characterization.
  • Site-specific biotinylation via AviTag for oriented immobilization.
  • Expressed in HEK293 cells for human-like glycosylation.
  • MALS-verified oligomeric state for consistent biophysical behavior.
  • High purity (>95% by SDS-PAGE) for reproducible assay results.
  • Lyophilized formulation with trehalose for enhanced stability; store at -20°C or lower.
Catalog No. 50-235-7362 Supplier ACROBiosystems Supplier No. RSFV82E725UG
Biotinylated prefusion F0 protein from Human respiratory syncytial virus A (strain A2), engineered with a C-terminal polyhistidine tag and an Avi tag for site-specific enzymatic biotinylation. The protein is expressed in HEK293 cells, MALS-verified, and supplied lyophilized (25 μg) for assay development, binding studies, and antibody characterization.
  • Site-specific biotinylation via AviTag for oriented immobilization.
  • Expressed in HEK293 cells for human-like glycosylation.
  • MALS-verified oligomeric state for consistent biophysical behavior.
  • High purity (>95% by SDS-PAGE) for reproducible assay results.
  • Lyophilized formulation with trehalose for enhanced stability; store at -20°C or lower.