Learn More
ACROBiosystems ACROBiosystems Biotinylated Human Siglec-2 / CD22 Protein, His,Avitag™ (MALS verified)
Catalog No. 502106609
Encompass_Preferred
Recombinant Protein;25UG;Biotinylated Human Siglec-2, His,Avitag (SI2-H82E3) is expressed from human 293 cells (HEK293). It contains AA Asp 20 - Arg 687 (Accession # P20273-1).;This protein carries a polyhistidine tag at the C-terminus, followed by an Avi tag. The protein has a calculated MW of 78.7 kDa. The protein migrates as 100-115 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Biotinylated Human Siglec-2, His,Avitag (Cat. No. SI2-H82E3) at 1 μg/mL (100 μL/well) on streptavidin (Cat. No. STN-N5116) precoated (0.5 μg/well) plate can bind Monoclonal Anti-Human Siglec-2 Antibody, Human IgG1 with a linear range of 1-10 ng/mL (QC tested).
Catalog No. 50-210-6609 Supplier ACROBiosystems Supplier No. SI2H82E325UG
Recombinant Protein;25UG;Biotinylated Human Siglec-2, His,Avitag (SI2-H82E3) is expressed from human 293 cells (HEK293). It contains AA Asp 20 - Arg 687 (Accession # P20273-1).;This protein carries a polyhistidine tag at the C-terminus, followed by an Avi tag. The protein has a calculated MW of 78.7 kDa. The protein migrates as 100-115 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Biotinylated Human Siglec-2, His,Avitag (Cat. No. SI2-H82E3) at 1 μg/mL (100 μL/well) on streptavidin (Cat. No. STN-N5116) precoated (0.5 μg/well) plate can bind Monoclonal Anti-Human Siglec-2 Antibody, Human IgG1 with a linear range of 1-10 ng/mL (QC tested).