Learn More
ACROBiosystems ACROBiosystems Biotinylated Human Siglec-3 / CD33 Protein, Fc,Avitag™
Catalog No. 502106887
Encompass_Preferred
Recombinant Protein;25UG;Biotinylated Human Siglec-3,Fc,Avitag™ (CD3-H82F3) is expressed from human 293 cells (HEK293). It contains AA Asp 18 - His 259 (Accession # AAH28152.1).;This protein carries a human IgG1 Fc tag at the C-terminus, followed by a Avi tag (Avitag™). The protein has a calculated MW of 54.9 kDa.
Catalog No. 50-210-6887 Supplier ACROBiosystems Supplier No. CD3H82F325UG
Recombinant Protein;25UG;Biotinylated Human Siglec-3,Fc,Avitag™ (CD3-H82F3) is expressed from human 293 cells (HEK293). It contains AA Asp 18 - His 259 (Accession # AAH28152.1).;This protein carries a human IgG1 Fc tag at the C-terminus, followed by a Avi tag (Avitag™). The protein has a calculated MW of 54.9 kDa.