Learn More
ACROBiosystems Biotinylated Human SIRP alphaV8 / CD172a Protein, His,Avitag™
Catalog No. 502055046
Encompass_Preferred
Recombinant Protein;25UG;Biotinylated Human SIRP alpha V8, His,Avitag (SI8-H82W6) is expressed from human 293 cells (HEK293). It contains AA Glu 31- Arg 369 (S44L, S50T, I52T, H54R, V57A) (Accession # ATD50864.1).;This protein carries a polyhistidine tag at the C-terminus, followed by an Avi tag. The protein has a calculated MW of 40.8 kDa. The protein migrates as 50-60 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Human CD47, Mouse IgG2a Fc Tag, low endotoxin (Cat. No. CD7-H52A5) at 2 μg/mL (100 μL/well) can bind Biotinylated Human SIRP alphaV8, His,Avitag (Cat. No. SI8-H82W6) with a linear range of 0.02-1.25 μg/mL (QC tested).
Catalog No. 50-205-5046 Supplier ACROBiosystems Supplier No. SI8H82W625UG
Recombinant Protein;25UG;Biotinylated Human SIRP alpha V8, His,Avitag (SI8-H82W6) is expressed from human 293 cells (HEK293). It contains AA Glu 31- Arg 369 (S44L, S50T, I52T, H54R, V57A) (Accession # ATD50864.1).;This protein carries a polyhistidine tag at the C-terminus, followed by an Avi tag. The protein has a calculated MW of 40.8 kDa. The protein migrates as 50-60 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Human CD47, Mouse IgG2a Fc Tag, low endotoxin (Cat. No. CD7-H52A5) at 2 μg/mL (100 μL/well) can bind Biotinylated Human SIRP alphaV8, His,Avitag (Cat. No. SI8-H82W6) with a linear range of 0.02-1.25 μg/mL (QC tested).