Learn More
ACROBiosystems Biotinylated Human SIRP alphaV2 / CD172a Protein, His,Avitag™
Catalog No. 502055044
Encompass_Preferred
Recombinant Protein;25UG;Biotinylated Human SIRP alphaV2, His,Avitag (SI2-H82W8) is expressed from human 293 cells (HEK293). It contains AA Glu 31 - Arg 369 (Accession # ATD50864.1).;This protein carries a polyhistidine tag at the C-terminus, followed by an Avi tag. The protein has a calculated MW of 40.9 kDa. The protein migrates as 55-66 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Human CD47, Fc Tag (HPLC-verified) (Cat. No. CD7-H5256) at 2 μg/mL (100 μL/well) can bind Biotinylated Human SIRP alphaV2, His,Avitag (Cat. No. SI2-H82W8) with a linear range of 0.039-0.625 μg/mL (QC tested).
Catalog No. 50-205-5044 Supplier ACROBiosystems Supplier No. SI2H82W825UG
Recombinant Protein;25UG;Biotinylated Human SIRP alphaV2, His,Avitag (SI2-H82W8) is expressed from human 293 cells (HEK293). It contains AA Glu 31 - Arg 369 (Accession # ATD50864.1).;This protein carries a polyhistidine tag at the C-terminus, followed by an Avi tag. The protein has a calculated MW of 40.9 kDa. The protein migrates as 55-66 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Human CD47, Fc Tag (HPLC-verified) (Cat. No. CD7-H5256) at 2 μg/mL (100 μL/well) can bind Biotinylated Human SIRP alphaV2, His,Avitag (Cat. No. SI2-H82W8) with a linear range of 0.039-0.625 μg/mL (QC tested).