Learn More
ACROBiosystems ACROBiosystems Biotinylated Human SLAMF1 / SLAM / CD150 Protein, His,Avitag™
Catalog No. 502106724
Encompass_Preferred
Recombinant Protein;25UG;Biotinylated Human SLAMF1, His,Avitag (SL1-H82E3) is expressed from human 293 cells (HEK293). It contains AA Ala 21 - Pro 237 (Accession # Q13291-1).;This protein carries a polyhistidine tag at the C-terminus, followed by an Avi tag. The protein has a calculated MW of 28.0 kDa. The protein migrates as 42-60 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.
Catalog No. 50-210-6724 Supplier ACROBiosystems Supplier No. SL1H82E325UG
Recombinant Protein;25UG;Biotinylated Human SLAMF1, His,Avitag (SL1-H82E3) is expressed from human 293 cells (HEK293). It contains AA Ala 21 - Pro 237 (Accession # Q13291-1).;This protein carries a polyhistidine tag at the C-terminus, followed by an Avi tag. The protein has a calculated MW of 28.0 kDa. The protein migrates as 42-60 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.