Learn More
ACROBiosystems Biotinylated Human Syndecan-1 Protein, His,Avitag™
Catalog No. 502055181
Encompass_Preferred
Recombinant Protein;200UG;Biotinylated Human Syndecan-1, His,Avitag™ (SY1-H82E2) is expressed from human 293 cells (HEK293). It contains AA Gln 23 - Gly 254 (Accession # P18827-1).;This protein carries a polyhistidine tag at the C-terminus, followed by an Avi tag. The protein has a calculated MW of 27.5 kDa. The protein migrates as 43-55 kDa under reducing (R) condition (SDS-PAGE) due to Glycosylation.;Immobilized Biotinylated Human Syndecan-1, His,Avitag (Cat. No. SY1-H82E2) at 1 μg/mL (100 μL/well) on streptavidin (Cat. No. STN-N5116) (0.5 μg/well) plate, can bind Anti-Syndecan-1 antibody, Mouse IgG1 with a linear range of 0.4-6 ng/mL (QC tested).;Immobilized Anti-Syndecan-1 antibody, Mouse IgG1 at 1 μg/mL (100 μL/well)onstreptavidin (Cat. No. STN-N5116) (0.1 μg/well) plate, can bind Biotinylated Human Syndecan-1, His,Avitag (Cat. No. SY1-H82E2) with a linear range of 2-63 ng/mL (Routinely tested).
Catalog No. 50-205-5181 Supplier ACROBiosystems Supplier No. SY1H82E2200UG
Recombinant Protein;200UG;Biotinylated Human Syndecan-1, His,Avitag™ (SY1-H82E2) is expressed from human 293 cells (HEK293). It contains AA Gln 23 - Gly 254 (Accession # P18827-1).;This protein carries a polyhistidine tag at the C-terminus, followed by an Avi tag. The protein has a calculated MW of 27.5 kDa. The protein migrates as 43-55 kDa under reducing (R) condition (SDS-PAGE) due to Glycosylation.;Immobilized Biotinylated Human Syndecan-1, His,Avitag (Cat. No. SY1-H82E2) at 1 μg/mL (100 μL/well) on streptavidin (Cat. No. STN-N5116) (0.5 μg/well) plate, can bind Anti-Syndecan-1 antibody, Mouse IgG1 with a linear range of 0.4-6 ng/mL (QC tested).;Immobilized Anti-Syndecan-1 antibody, Mouse IgG1 at 1 μg/mL (100 μL/well)onstreptavidin (Cat. No. STN-N5116) (0.1 μg/well) plate, can bind Biotinylated Human Syndecan-1, His,Avitag (Cat. No. SY1-H82E2) with a linear range of 2-63 ng/mL (Routinely tested).