Learn More
ACROBiosystems Biotinylated Human Syndecan-1 Protein, Fc,Avitag™
Catalog No. 502055179
Encompass_Preferred
Recombinant Protein;200UG;Biotinylated Human Syndecan-1, Fc,Avitag (SY1-H82F3) is expressed from human 293 cells (HEK293). It contains AA Gln 23 - Gly 254 (Accession # P18827-1).;This protein carries a human IgG1 Fc tag at the C-terminus, followed by a Avi tag (Avitag™). The protein has a calculated MW of 52.0 kDa. The protein migrates as 60-66 kDa under reducing (R) condition (SDS-PAGE) due to Glycosylation.;Immobilized Biotinylated Human Syndecan-1, Fc,Avitag (Cat. No. SY1-H82F3) at 1 μg/mL (100 μL/well) on streptavidin (Cat. No. STN-N5116) (0.5 μg/well) plate. can bind Anti-Syndecan-1 antibody, Mouse IgG1 with a linear range of 0.2-6 ng/mL (QC tested).;Immobilized Anti-Syndecan-1 antibody, Mouse IgG1 at 1 μg/mL (100 μL/well) can bind Biotinylated Human Syndecan-1, Fc,Avitag (Cat. No. SY1-H82F3) with a linear range of 0.5-8 ng/mL (Routinely tested).
Catalog No. 50-205-5179 Supplier ACROBiosystems Supplier No. SY1H82F3200UG
Recombinant Protein;200UG;Biotinylated Human Syndecan-1, Fc,Avitag (SY1-H82F3) is expressed from human 293 cells (HEK293). It contains AA Gln 23 - Gly 254 (Accession # P18827-1).;This protein carries a human IgG1 Fc tag at the C-terminus, followed by a Avi tag (Avitag™). The protein has a calculated MW of 52.0 kDa. The protein migrates as 60-66 kDa under reducing (R) condition (SDS-PAGE) due to Glycosylation.;Immobilized Biotinylated Human Syndecan-1, Fc,Avitag (Cat. No. SY1-H82F3) at 1 μg/mL (100 μL/well) on streptavidin (Cat. No. STN-N5116) (0.5 μg/well) plate. can bind Anti-Syndecan-1 antibody, Mouse IgG1 with a linear range of 0.2-6 ng/mL (QC tested).;Immobilized Anti-Syndecan-1 antibody, Mouse IgG1 at 1 μg/mL (100 μL/well) can bind Biotinylated Human Syndecan-1, Fc,Avitag (Cat. No. SY1-H82F3) with a linear range of 0.5-8 ng/mL (Routinely tested).