Learn More
Kactus Biotinylated Human TGF-beta RII/TGFBR2 Protein (TGF-HM3R2B) 100ug
Catalog No. NC3803965
Encompass
Recombinant Biotinylated Human TGF-beta RII/TGFBR2 Protein is expressed from HEK293 with mFc (IgG1) tag and Avi tag at C-Terminus.It contains Ile24-Asp159.
Catalog No. NC3803965 Supplier Kactus Supplier No. TGFHM3R2B
Recombinant Biotinylated Human TGF-beta RII/TGFBR2 Protein is expressed from HEK293 with mFc (IgG1) tag and Avi tag at C-Terminus.It contains Ile24-Asp159.