Learn More
ACROBiosystems Biotinylated Human TREM2 Protein, His,Avitag™
Catalog No. 502055198
Encompass_Preferred
Recombinant Protein;25UG;Biotinylated Human TREM2, His,Avitag™ (TR2-H82E7) is expressed from human 293 cells (HEK293). It contains AA His 19 - Ser 174 (Accession # Q9NZC2-1).;This protein carries a polyhistidine tag at the C-terminus, followed by an Avi tag. The protein has a calculated MW of 21.0 kDa. The protein migrates as 25-35 kDa under reducing (R) condition (SDS-PAGE) due to Glycosylation.;Immobilized Biotinylated Human TREM2, His,Avitag (Cat. No. TR2-H82E7) at 1 μg/mL (100 μL/well) on streptavidin (Cat. No. STN-N5116) precoated (2 μg/well) plate can bind Anti-TREM2 Antibody, Human IgG1 with a linear range of 0.2-2 ng/mL (QC tested).
Catalog No. 50-205-5198 Supplier ACROBiosystems Supplier No. TR2H82E725UG
Recombinant Protein;25UG;Biotinylated Human TREM2, His,Avitag™ (TR2-H82E7) is expressed from human 293 cells (HEK293). It contains AA His 19 - Ser 174 (Accession # Q9NZC2-1).;This protein carries a polyhistidine tag at the C-terminus, followed by an Avi tag. The protein has a calculated MW of 21.0 kDa. The protein migrates as 25-35 kDa under reducing (R) condition (SDS-PAGE) due to Glycosylation.;Immobilized Biotinylated Human TREM2, His,Avitag (Cat. No. TR2-H82E7) at 1 μg/mL (100 μL/well) on streptavidin (Cat. No. STN-N5116) precoated (2 μg/well) plate can bind Anti-TREM2 Antibody, Human IgG1 with a linear range of 0.2-2 ng/mL (QC tested).