Learn More
ACROBiosystems Biotinylated Human TROP-2 (TACSTD2) Protein, C-terminal His and Avi tag, 25 μg
Supplier: ACROBiosystems TR2H82EA25UG
Biotinylated recombinant human TROP-2 (TACSTD2) ectodomain (residues 88-274) supplied with a C-terminal polyhistidine tag and Avi tag. Enzymatically biotinylated for site-specific labeling and verified by SEC-MALS for size and purity. Supplied lyophilized for long-term storage.
- Expressed in HEK293 cells.
- Residues Thr 88-Thr 274.
- Contains C-terminal polyhistidine tag and Avi tag.
- Site-specific enzymatic biotinylation via Avi tag.
- Purity: >90% by SDS-PAGE; >85% by SEC-MALS.
- Calculated molecular weight: 25.1 kDa; migrates ~35-45 kDa on SDS-PAGE.
- Format: lyophilized; available as 25 μg.
- Storage: lyophilized at ≤-20 °C; reconstituted storage at -70 °C under sterile conditions.
- Application: binding assays and detection (e.g., ELISA).
- Verification: SEC-MALS verified.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.