Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Biotinylated Isoxazole

Catalog No. 9005721MG Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
90-057-21MG 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 90-057-21MG Supplier Sigma Aldrich Supplier No. 9005721MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

When exposed to cells or tissue lysates, biotinylated isoxazole (b-isox) was shown to precipitate RNA-binding proteins with significant overlap to the components of RNA granules. By reversibly aggregating and disaggregating structures similar to RNA granules, b-isox has lent insight to polypeptides necessary for the reversible aggregation of RNA-binding proteins.This product was previously listed as T511617.Cell-free Formation of RNA Granules: Low Complexity Sequence Domains Form Dynamic Fibers within Hydrogels

Specifications

CAS 158923-09-2
Molecular Formula C36H44FeP2
Linear Formula C36H44FeP2
MDL Number MFCD00800280
Quantity 1 g
Synonym (1R)-1-(Dicyclohexylphosphino)-2-[(1R)-1-(diphenylphosphino)ethyl]ferrocene (acc to CAS); Josiphos SL-J004-1
Molecular Weight (g/mol) 594.53 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.