Learn More
ACROBiosystems Biotinylated Monoclonal Anti-FMC63 scFv Antibody, Mouse IgG1, Avitag™ (Y45)
Catalog No. 502054869
Encompass_Preferred
Antibody;200TESTS;IgG1/kappa;Specifically recognizes the antigen-recognition domain of FMC63 derived CARs.;Biotinylated Monoclonal Anti-FMC63 scFv Antibody, Mouse IgG1, Avitag (Cat. No. FM3-BY54) is expressed from human 293 cells.; ;2e5 of Anti-CD19 CAR-293 cells were stained with 100 μL of 1:50 dilution (2 μL stock solution in 100 μL FACS buffer) Biotinylated Monoclonal Anti-FMC63 scFv Antibody, Mouse IgG1, Avitag (Cat. No. FM3-BY54) and isotype control respectively, washed and then followed by PE-SA and analyzed with FACS (QC tested).
Catalog No. 50-205-4869 Supplier ACROBiosystems Supplier No. FM3BY54200TESTS
Antibody;200TESTS;IgG1/kappa;Specifically recognizes the antigen-recognition domain of FMC63 derived CARs.;Biotinylated Monoclonal Anti-FMC63 scFv Antibody, Mouse IgG1, Avitag (Cat. No. FM3-BY54) is expressed from human 293 cells.; ;2e5 of Anti-CD19 CAR-293 cells were stained with 100 μL of 1:50 dilution (2 μL stock solution in 100 μL FACS buffer) Biotinylated Monoclonal Anti-FMC63 scFv Antibody, Mouse IgG1, Avitag (Cat. No. FM3-BY54) and isotype control respectively, washed and then followed by PE-SA and analyzed with FACS (QC tested).