Learn More
ACROBiosystems ACROBiosystems Biotinylated Mouse BCMA / TNFRSF17 Protein, Fc,Avitag™
Catalog No. 502106459
Encompass_Preferred
Recombinant Protein;200UG;Biotinylated Mouse BCMA, Fc,Avitag (BCA-M82F0) is expressed from human 293 cells (HEK293). It contains AA Met 1 - Thr 49 (Accession # O88472-1).;This protein carries a human IgG1 Fc tag at the C-terminus, followed by a Avi tag (Avitag™). The protein has a calculated MW of 34.4 kDa. The protein migrates as 38-50 kDa under reducing (R) condition (SDS-PAGE) due to O-linked glycosylation which has been verified by O-Glycosidase and Neuraminidase digestion.;Immobilized Mouse BAFF, Mouse IgG2a Fc Tag (Cat. No. BAF-M5257) at 5 μg/mL (100 μL/well) can bind Biotinylated Mouse BCMA, Fc,Avitag (Cat. No. BCA-M82F0) with a linear range of 2-39 ng/mL (QC tested).;Immobilized Biotinylated Mouse BCMA, Fc,Avitag (Cat. No. BCA-M82F0) at 1 μg/mL (100 μL/well) on Recombinant Streptavidin (Cat. No. STN-N5116) precoated (0.5μg/well) plate, can bind Human APRIL, His,Flag Tag, active trimer (Cat. No. APL-H52D1) with a linear range of 10-78 ng/mL (Routinely tested).
Catalog No. 50-210-6459 Supplier ACROBiosystems Supplier No. BCAM82F0200UG
Recombinant Protein;200UG;Biotinylated Mouse BCMA, Fc,Avitag (BCA-M82F0) is expressed from human 293 cells (HEK293). It contains AA Met 1 - Thr 49 (Accession # O88472-1).;This protein carries a human IgG1 Fc tag at the C-terminus, followed by a Avi tag (Avitag™). The protein has a calculated MW of 34.4 kDa. The protein migrates as 38-50 kDa under reducing (R) condition (SDS-PAGE) due to O-linked glycosylation which has been verified by O-Glycosidase and Neuraminidase digestion.;Immobilized Mouse BAFF, Mouse IgG2a Fc Tag (Cat. No. BAF-M5257) at 5 μg/mL (100 μL/well) can bind Biotinylated Mouse BCMA, Fc,Avitag (Cat. No. BCA-M82F0) with a linear range of 2-39 ng/mL (QC tested).;Immobilized Biotinylated Mouse BCMA, Fc,Avitag (Cat. No. BCA-M82F0) at 1 μg/mL (100 μL/well) on Recombinant Streptavidin (Cat. No. STN-N5116) precoated (0.5μg/well) plate, can bind Human APRIL, His,Flag Tag, active trimer (Cat. No. APL-H52D1) with a linear range of 10-78 ng/mL (Routinely tested).