Learn More
ACROBiosystems ACROBiosystems Biotinylated Mouse CD200 / OX-2 Protein, Fc,Avitag™ (MALS verified)
Catalog No. 502106432
Encompass_Preferred
Recombinant Protein;25UG;Biotinylated Mouse CD200, Fc,Avitag (OX2-M82F3) is expressed from human 293 cells (HEK293). It contains AA Gln 31 - Gly 232 (Accession # O54901-1).;This protein carries a human IgG1 Fc tag at the C-terminus, followed by a Avi tag (Avitag™). The protein has a calculated MW of 50.5 kDa. The protein migrates as 66-75 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Human CD200 R1, His Tag (Cat. No. CR2-H52H6) at 5 μg/mL (100 μL/well) can bind Biotinylated Mouse CD200, Fc,Avitag (Cat. No. OX2-M82F3) with a linear range of 0.02-0.5 μg/mL (QC tested).
Catalog No. 50-210-6432 Supplier ACROBiosystems Supplier No. OX2M82F325UG
Recombinant Protein;25UG;Biotinylated Mouse CD200, Fc,Avitag (OX2-M82F3) is expressed from human 293 cells (HEK293). It contains AA Gln 31 - Gly 232 (Accession # O54901-1).;This protein carries a human IgG1 Fc tag at the C-terminus, followed by a Avi tag (Avitag™). The protein has a calculated MW of 50.5 kDa. The protein migrates as 66-75 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Human CD200 R1, His Tag (Cat. No. CR2-H52H6) at 5 μg/mL (100 μL/well) can bind Biotinylated Mouse CD200, Fc,Avitag (Cat. No. OX2-M82F3) with a linear range of 0.02-0.5 μg/mL (QC tested).