Learn More
ACROBiosystems ACROBiosystems Biotinylated Mouse CD40 / TNFRSF5 Protein, Fc,Avitag™ (MALS verified)
Supplier:  ACROBiosystems CD0M82F3200UG
Encompass_Preferred
Recombinant Protein;200UG;Biotinylated Mouse CD40, Fc,Avitag (CD0-M82F3) is expressed from human 293 cells (HEK293). It contains AA Val 24 - Arg 193 (Accession # P27512-1).;This protein carries a human IgG1 Fc tag at the C-terminus, followed by a Avi tag (Avitag™). The protein has a calculated MW of 47.1 kDa. The protein migrates as 55-60 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Biotinylated Mouse CD40, Fc,Avitag (Cat. No. CD0-M82F3) at 1 μg/mL (100 μL/well) on streptavidin (Cat. No. STN-N5116) precoated (0.5 μg/well) plate can bind Mouse CD40 Ligand, His Tag (active trimer) (Cat. No. CDL-M5248) with a linear range of 10-78 ng/mL (QC tested).
Catalog No. 50-210-6466