Learn More
ACROBiosystems ACROBiosystems Biotinylated Mouse Fc gamma RI / CD64 Protein, His,Avitag™ (MALS verified)
Catalog No. 502106221
Encompass_Preferred
Recombinant Protein;25UG;Biotinylated Mouse CD64, His,Avitag (CD4-M82E7) is expressed from human 293 cells (HEK293). It contains AA Glu 25 - Pro 297 (Accession # P26151-1).;This protein carries a polyhistidine tag at the C-terminus, followed by an Avi tag. The protein has a calculated MW of 34.0 kDa. The protein migrates as 45-60 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Biotinylated Mouse CD64, His,Avitag (Cat. No. CD4-M82E7) captured on Biotin CAP - Series S sensor Chip can bind Herceptin with an affinity constant of 59.8 nM as determined in a SPR assay (Biacore T200) (QC tested).
Catalog No. 50-210-6221 Supplier ACROBiosystems Supplier No. CD4M82E725UG
Recombinant Protein;25UG;Biotinylated Mouse CD64, His,Avitag (CD4-M82E7) is expressed from human 293 cells (HEK293). It contains AA Glu 25 - Pro 297 (Accession # P26151-1).;This protein carries a polyhistidine tag at the C-terminus, followed by an Avi tag. The protein has a calculated MW of 34.0 kDa. The protein migrates as 45-60 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Biotinylated Mouse CD64, His,Avitag (Cat. No. CD4-M82E7) captured on Biotin CAP - Series S sensor Chip can bind Herceptin with an affinity constant of 59.8 nM as determined in a SPR assay (Biacore T200) (QC tested).