Learn More
ACROBiosystems ACROBiosystems Biotinylated Mouse FOLR1 Protein, His,Avitag™
Catalog No. 502106643
Encompass_Preferred
Recombinant Protein;25UG;Biotinylated Mouse FOLR1, His,Avitag (FO1-M82E9) is expressed from human 293 cells (HEK293). It contains AA Thr 25 - Ser 232 (Accession # P35846-1).;This protein carries a polyhistidine tag at the C-terminus, followed by an Avi tag (Avitag™). The protein has a calculated MW of 26.9 kDa. The protein migrates as 32-45 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Folic acid-BSA at 5 μg/mL (100 μL/well) can bind Biotinylated Mouse FOLR1, His,Avitag (Cat. No. FO1-M82E9) with a linear range of 0.008-0.222 μg/mL (QC tested).
Catalog No. 50-210-6643 Supplier ACROBiosystems Supplier No. FO1M82E925UG
Recombinant Protein;25UG;Biotinylated Mouse FOLR1, His,Avitag (FO1-M82E9) is expressed from human 293 cells (HEK293). It contains AA Thr 25 - Ser 232 (Accession # P35846-1).;This protein carries a polyhistidine tag at the C-terminus, followed by an Avi tag (Avitag™). The protein has a calculated MW of 26.9 kDa. The protein migrates as 32-45 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Folic acid-BSA at 5 μg/mL (100 μL/well) can bind Biotinylated Mouse FOLR1, His,Avitag (Cat. No. FO1-M82E9) with a linear range of 0.008-0.222 μg/mL (QC tested).