Learn More
ACROBiosystems Biotinylated Mouse IgG1 Fc protein, Avitag™
Catalog No. 502055006
Encompass_Preferred
Recombinant Protein;25UG;Biotinylated Mouse IgG1 Fc, Avitag™ (IG1-M8211) is expressed from human 293 cells (HEK293). It contains AA Val 98 - Lys 324 (Accession # AAK53870.1).;This protein carries an Avi tag at the C-terminus. The protein has a calculated MW of 27.4 kDa. The protein migrates as 33 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.
Catalog No. 50-205-5006 Supplier ACROBiosystems Supplier No. IG1M821125UG
Recombinant Protein;25UG;Biotinylated Mouse IgG1 Fc, Avitag™ (IG1-M8211) is expressed from human 293 cells (HEK293). It contains AA Val 98 - Lys 324 (Accession # AAK53870.1).;This protein carries an Avi tag at the C-terminus. The protein has a calculated MW of 27.4 kDa. The protein migrates as 33 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.