Learn More
ACROBiosystems Biotinylated Mouse IL-6 R alpha / CD126 Protein, His,Avitag™
Catalog No. 502055136
Encompass_Preferred
Recombinant Protein;200UG;Biotinylated Mouse IL-6 R alpha, His,Avitag (ILR-M82E9) is expressed from human 293 cells (HEK293). It contains AA Leu 20 - Glu 357 (Accession # P22272-1).;This protein carries a polyhistidine tag at the C-terminus, followed by an Avi tag. The protein has a calculated MW of 41.0 kDa. The protein migrates as 45-66 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Mouse Human IL-6, His Tag (Cat. No. IL6-M5245) at 5 μg/mL (100 μL/well) can bind Biotinylated Mouse IL-6 R alpha, His,Avitag (Cat. No. ILR-M82E9) with a linear range of 20-156 ng/mL (QC tested).
Catalog No. 50-205-5136 Supplier ACROBiosystems Supplier No. ILRM82E9200UG
Recombinant Protein;200UG;Biotinylated Mouse IL-6 R alpha, His,Avitag (ILR-M82E9) is expressed from human 293 cells (HEK293). It contains AA Leu 20 - Glu 357 (Accession # P22272-1).;This protein carries a polyhistidine tag at the C-terminus, followed by an Avi tag. The protein has a calculated MW of 41.0 kDa. The protein migrates as 45-66 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Mouse Human IL-6, His Tag (Cat. No. IL6-M5245) at 5 μg/mL (100 μL/well) can bind Biotinylated Mouse IL-6 R alpha, His,Avitag (Cat. No. ILR-M82E9) with a linear range of 20-156 ng/mL (QC tested).