Learn More
ACROBiosystems ACROBiosystems Biotinylated Mouse Mesothelin / MSLN Protein, His,Avitag™
Catalog No. 502106263
Encompass_Preferred
Recombinant Protein;25UG;Biotinylated Mouse Mesothelin, His,Avitag (MSN-M82E7) is expressed from human 293 cells (HEK293). It contains AA Asp 298 - Ser 600 (Accession # Q61468-1).;This protein carries a polyhistidine tag at the C-terminus, followed by an Avi tag. The protein has a calculated MW of 37.7 kDa. The protein migrates as 45-60 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Biotinylated Mouse Mesothelin, His,Avitag (Cat. No. MSN-M82E7) at 1 μg/mL (100 μL/well) on streptavidin (Cat. No. STN-N5116) precoated (0.5 μg/well) plate can bind Monoclonal Anti-Human MSLN Antibody, Human IgG1 with a linear range of 10-78 ng/mL (QC tested).
Catalog No. 50-210-6263 Supplier ACROBiosystems Supplier No. MSNM82E725UG
Recombinant Protein;25UG;Biotinylated Mouse Mesothelin, His,Avitag (MSN-M82E7) is expressed from human 293 cells (HEK293). It contains AA Asp 298 - Ser 600 (Accession # Q61468-1).;This protein carries a polyhistidine tag at the C-terminus, followed by an Avi tag. The protein has a calculated MW of 37.7 kDa. The protein migrates as 45-60 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Biotinylated Mouse Mesothelin, His,Avitag (Cat. No. MSN-M82E7) at 1 μg/mL (100 μL/well) on streptavidin (Cat. No. STN-N5116) precoated (0.5 μg/well) plate can bind Monoclonal Anti-Human MSLN Antibody, Human IgG1 with a linear range of 10-78 ng/mL (QC tested).