Learn More
ACROBiosystems ACROBiosystems Biotinylated Mouse NKp46 / NCR1 / CD335 Protein, His,Avitag™ (MALS verified)
Catalog No. 502106949
Encompass_Preferred
Recombinant Protein;200UG;Biotinylated Mouse NKp46, His,Avitag™ (NC1-M82H5) is expressed from human 293 cells (HEK293). It contains AA Glu 22 - Asn 255 (Accession # Q8C567-1).;This protein carries a polyhistidine tag at the C-terminus, followed by an Avi tag. The protein has a calculated MW of 19.6 kDa.
Catalog No. 50-210-6949 Supplier ACROBiosystems Supplier No. NC1M82H5200UG
Recombinant Protein;200UG;Biotinylated Mouse NKp46, His,Avitag™ (NC1-M82H5) is expressed from human 293 cells (HEK293). It contains AA Glu 22 - Asn 255 (Accession # Q8C567-1).;This protein carries a polyhistidine tag at the C-terminus, followed by an Avi tag. The protein has a calculated MW of 19.6 kDa.