Learn More
ACROBiosystems ACROBiosystems Biotinylated Mouse OX40 / TNFRSF4 / CD134 Protein, His,Avitag™
Catalog No. 502106668
Encompass_Preferred
Recombinant Protein;200UG;Biotinylated Mouse OX40, His,Avitag (OX0-M82E5) is expressed from human 293 cells (HEK293). It contains AA Val 20 - Pro 211 (Accession # P47741-1).;This protein carries a polyhistidine tag at the C-terminus, followed by an Avi tag. The protein has a calculated MW of 25.0 kDa. The protein migrates as 35-50 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Mouse OX40 Ligand, His Tag (Cat. No. TN4-M5241) at 2 μg/mL (100 μL/well) can bind Biotinylated Mouse OX40, His,Avitag (Cat. No. OX0-M82E5) with a linear range of 0.6-10 ng/mL (QC tested).
Catalog No. 50-210-6668 Supplier ACROBiosystems Supplier No. OX0M82E5200UG
Recombinant Protein;200UG;Biotinylated Mouse OX40, His,Avitag (OX0-M82E5) is expressed from human 293 cells (HEK293). It contains AA Val 20 - Pro 211 (Accession # P47741-1).;This protein carries a polyhistidine tag at the C-terminus, followed by an Avi tag. The protein has a calculated MW of 25.0 kDa. The protein migrates as 35-50 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Mouse OX40 Ligand, His Tag (Cat. No. TN4-M5241) at 2 μg/mL (100 μL/well) can bind Biotinylated Mouse OX40, His,Avitag (Cat. No. OX0-M82E5) with a linear range of 0.6-10 ng/mL (QC tested).