Learn More
ACROBiosystems ACROBiosystems Biotinylated Mouse Serum Albumin Protein, His,Avitag™ (MALS verified)
Catalog No. 502106833
Encompass_Preferred
Recombinant Protein;200UG;Biotinylated Mouse Serum Albumin, His,Avitag (MSA-M82E4) is expressed from human 293 cells (HEK293). It contains AA Glu 25 - Ala 608 (Accession # P07724-1).;This protein carries a polyhistidine tag at the C-terminus, followed by an Avi tag. The protein has a calculated MW of 69.5 kDa. The protein migrates as 66-80 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.
Catalog No. 50-210-6833 Supplier ACROBiosystems Supplier No. MSAM82E4200UG
Recombinant Protein;200UG;Biotinylated Mouse Serum Albumin, His,Avitag (MSA-M82E4) is expressed from human 293 cells (HEK293). It contains AA Glu 25 - Ala 608 (Accession # P07724-1).;This protein carries a polyhistidine tag at the C-terminus, followed by an Avi tag. The protein has a calculated MW of 69.5 kDa. The protein migrates as 66-80 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.