Learn More
ACROBiosystems Biotinylated Mouse TGF-beta RII / TGFBR2 Protein, His,Avitag™
Catalog No. 502055079
Encompass_Preferred
Recombinant Protein;25UG;Biotinylated Mouse TGF-beta RII, His,Avitag™ (TG2-M82H5) is expressed from human 293 cells (HEK293). It contains AA Ile 24 - Asp 184 (Accession # Q62312-1).;This protein carries a polyhistidine tag at the C-terminus, followed by an Avi tag. The protein has a calculated MW of 21.7 kDa. The protein migrates as 32-45 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Human Latent TGFB1, His Tag (Cat. No. TG1-H524x) at 1 μg/mL (100 μL/well) can bind Biotinylated Mouse TGF-beta RII, His,Avitag (Cat. No. TG2-M82H5) with a linear range of 1-31 ng/mL (QC tested).
Catalog No. 50-205-5079 Supplier ACROBiosystems Supplier No. TG2M82H525UG
Recombinant Protein;25UG;Biotinylated Mouse TGF-beta RII, His,Avitag™ (TG2-M82H5) is expressed from human 293 cells (HEK293). It contains AA Ile 24 - Asp 184 (Accession # Q62312-1).;This protein carries a polyhistidine tag at the C-terminus, followed by an Avi tag. The protein has a calculated MW of 21.7 kDa. The protein migrates as 32-45 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Human Latent TGFB1, His Tag (Cat. No. TG1-H524x) at 1 μg/mL (100 μL/well) can bind Biotinylated Mouse TGF-beta RII, His,Avitag (Cat. No. TG2-M82H5) with a linear range of 1-31 ng/mL (QC tested).