Learn More
ACROBiosystems ACROBiosystems Biotinylated Mouse TSLP Protein, His,Avitag™
Catalog No. 502106874
Encompass_Preferred
Recombinant Protein;200UG;Biotinylated Mouse TSLP, His,Avitag™ (TSP-M82E3) is expressed from human 293 cells (HEK293). It contains AA Tyr 20 - Glu 140 (Accession # Q9JIE6-1).;This protein carries a polyhistidine tag at the C-terminus, followed by an Avi tag. The protein has a calculated MW of 17.6 kDa.
Catalog No. 50-210-6874 Supplier ACROBiosystems Supplier No. TSPM82E3200UG
Recombinant Protein;200UG;Biotinylated Mouse TSLP, His,Avitag™ (TSP-M82E3) is expressed from human 293 cells (HEK293). It contains AA Tyr 20 - Glu 140 (Accession # Q9JIE6-1).;This protein carries a polyhistidine tag at the C-terminus, followed by an Avi tag. The protein has a calculated MW of 17.6 kDa.