Learn More
ACROBiosystems ACROBiosystems Biotinylated Mouse Fc gamma RIII / CD16 Protein, His,Avitag™ (MALS verified)
Catalog No. 502106218
Encompass_Preferred
Recombinant Protein;25UG;Biotinylated Mouse CD16, His,Avitag (FC6-M82E0) is expressed from human 293 cells (HEK293). It contains AA Leu 32 - Thr 215 (Accession # Q5D5I8-1).;This protein carries a polyhistidine tag at the C-terminus, followed by an Avi tag. The protein has a calculated MW of 24.6 kDa. The protein migrates as 35-50 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Biotinylated Mouse CD16, His,Avitag (Cat. No. FC6-M82E0) captured on Biotin CAP- Series S Sensor Chip can bind Monoclonal Anti-Human CD3 Antibody, Mouse IgG2a (Clone: OKT3) with an affinity constant of 0.345 μM as determined in a SPR assay (Biacore T200) (QC tested).
Catalog No. 50-210-6218 Supplier ACROBiosystems Supplier No. FC6M82E025UG
Recombinant Protein;25UG;Biotinylated Mouse CD16, His,Avitag (FC6-M82E0) is expressed from human 293 cells (HEK293). It contains AA Leu 32 - Thr 215 (Accession # Q5D5I8-1).;This protein carries a polyhistidine tag at the C-terminus, followed by an Avi tag. The protein has a calculated MW of 24.6 kDa. The protein migrates as 35-50 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Biotinylated Mouse CD16, His,Avitag (Cat. No. FC6-M82E0) captured on Biotin CAP- Series S Sensor Chip can bind Monoclonal Anti-Human CD3 Antibody, Mouse IgG2a (Clone: OKT3) with an affinity constant of 0.345 μM as determined in a SPR assay (Biacore T200) (QC tested).