Learn More
ACROBiosystems Biotinylated Onchocerca volvulus OV16 protein, Avi,GST Tag
Catalog No. 502055243
Encompass_Preferred
Recombinant Protein;200UG;Biotinylated Onchocerca volvulus OV16 protein, Avi,GST Tag (OV6-V81G9) is expressed from E.coli cells. It contains AA Lys 17 - Asp 197 (Accession # P31729-1).;This protein carries an Avi tag at the N-terminus, followed by a GST tag. The protein has a calculated MW of 48.6 kDa. The protein migrates as 46-50 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.
Catalog No. 50-205-5243 Supplier ACROBiosystems Supplier No. OV6V81G9200UG
Recombinant Protein;200UG;Biotinylated Onchocerca volvulus OV16 protein, Avi,GST Tag (OV6-V81G9) is expressed from E.coli cells. It contains AA Lys 17 - Asp 197 (Accession # P31729-1).;This protein carries an Avi tag at the N-terminus, followed by a GST tag. The protein has a calculated MW of 48.6 kDa. The protein migrates as 46-50 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.