Learn More
ACROBiosystems ACROBiosystems Biotinylated SARS-CoV-2 S protein RBD (N501Y), His,Avitag™ (MALS verified)
Catalog No. 502107316
Encompass_Preferred
Recombinant Protein;25UG;Biotinylated SARS-CoV-2 S protein RBD (N501Y), His,Avitag (SPD-C82E6) is expressed from human 293 cells (HEK293). It contains AA Arg 319 - Lys 537 (Accession # QHD43416.1 (N501Y)).;This protein carries a polyhistidine tag at the C-terminus, followed by an Avi tag. The protein has a calculated MW of 28.3 kDa. The protein migrates as 33-40 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Human ACE2, Fc Tag (Cat. No. AC2-H5257) at 1 μg/mL (100 μL/well) can bind Biotinylated SARS-CoV-2 S protein RBD (N501Y), His,Avitag (Cat. No. SPD-C82E6) with a linear range of 2-39 ng/mL (QC tested).;Immobilized Biotinylated SARS-CoV-2 S protein RBD (N501Y), His,Avitag (Cat. No. SPD-C82E6) at 1 μg/mL (100 μL/well) on streptavidin (Cat. No. STN-N5116) precoated (0.5 μg/well) plate can bind Human ACE2, Fc Tag (Cat. No. AC2-H5257) with a linear range of 0.1-2 ng/mL (Routinely tested).
Catalog No. 50-210-7316 Supplier ACROBiosystems Supplier No. SPDC82E625UG
Recombinant Protein;25UG;Biotinylated SARS-CoV-2 S protein RBD (N501Y), His,Avitag (SPD-C82E6) is expressed from human 293 cells (HEK293). It contains AA Arg 319 - Lys 537 (Accession # QHD43416.1 (N501Y)).;This protein carries a polyhistidine tag at the C-terminus, followed by an Avi tag. The protein has a calculated MW of 28.3 kDa. The protein migrates as 33-40 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Human ACE2, Fc Tag (Cat. No. AC2-H5257) at 1 μg/mL (100 μL/well) can bind Biotinylated SARS-CoV-2 S protein RBD (N501Y), His,Avitag (Cat. No. SPD-C82E6) with a linear range of 2-39 ng/mL (QC tested).;Immobilized Biotinylated SARS-CoV-2 S protein RBD (N501Y), His,Avitag (Cat. No. SPD-C82E6) at 1 μg/mL (100 μL/well) on streptavidin (Cat. No. STN-N5116) precoated (0.5 μg/well) plate can bind Human ACE2, Fc Tag (Cat. No. AC2-H5257) with a linear range of 0.1-2 ng/mL (Routinely tested).