Learn More
ACROBiosystems ACROBiosystems Biotinylated SARS-CoV-2 S protein (L18F, T20N, P26S, D138Y, R190S, K417T, E484K, N501Y, D614G, H655Y, T1027I, V1176F) trimer, His,Avitag™ (MALS verified)
Catalog No. 502107325
Encompass_Preferred
Recombinant Protein;200UG;Biotinylated SARS-CoV-2 S protein trimer, His,Avitag (SPN-C82E6) is expressed from human 293 cells (HEK293). It contains AA Val 16 - Pro 1213 (Accession # QHD43416.1). The recombinant protein is expressed from human 293 cells (HEK293) with T4 fibritin trimerization motif and a polyhistidine tag at the C-terminus. Proline substitutions (F817P/ A892P/ A899P/ A942P/ K986P/ V987P) and alanine substitutions (R683A and R685A) are introduced to stabilize the trimeric prefusion state of SARS-CoV-2 S protein and abolish the furin cleavage site, respectively. L18F/ T20N/ P26S/ D138Y/ R190S/ K417T/ E484K/ N501Y/ D614G/ H655Y/ T1027I/ V1176F) mutations were identified on the spike protein in the SARS-CoV-2 variant (known as P.1) which emerged in Brazil.;This protein carries a polyhistidine tag at the C-terminus, followed by an Avi tag. The protein has a calculated MW of 139.8 kDa. The protein migrates as 160-200 kDa under reducing (R) condition (SDS-PAGE) due to glycosyla
Catalog No. 50-210-7325 Supplier ACROBiosystems Supplier No. SPNC82E6200UG
Recombinant Protein;200UG;Biotinylated SARS-CoV-2 S protein trimer, His,Avitag (SPN-C82E6) is expressed from human 293 cells (HEK293). It contains AA Val 16 - Pro 1213 (Accession # QHD43416.1). The recombinant protein is expressed from human 293 cells (HEK293) with T4 fibritin trimerization motif and a polyhistidine tag at the C-terminus. Proline substitutions (F817P/ A892P/ A899P/ A942P/ K986P/ V987P) and alanine substitutions (R683A and R685A) are introduced to stabilize the trimeric prefusion state of SARS-CoV-2 S protein and abolish the furin cleavage site, respectively. L18F/ T20N/ P26S/ D138Y/ R190S/ K417T/ E484K/ N501Y/ D614G/ H655Y/ T1027I/ V1176F) mutations were identified on the spike protein in the SARS-CoV-2 variant (known as P.1) which emerged in Brazil.;This protein carries a polyhistidine tag at the C-terminus, followed by an Avi tag. The protein has a calculated MW of 139.8 kDa. The protein migrates as 160-200 kDa under reducing (R) condition (SDS-PAGE) due to glycosyla