Learn More
ACROBiosystems ACROBiosystems Biotinylated SARS-CoV-2 S protein (HV69-70del, Y144del, N501Y, A570D, D614G, P681H, T716I, S982A, D1118H) trimer, His,Avitag™ (MALS verified)
Catalog No. 502107327
Encompass_Preferred
Recombinant Protein;200UG;Biotinylated SARS-CoV-2 S protein trimer, His,Avitag (SPN-C82E5) is expressed from human 293 cells (HEK293). It contains AA Val 16 - Pro 1213 (Accession # QHD43416.1). The recombinant protein is expressed from human 293 cells (HEK293) with T4 fibritin trimerization motif and a polyhistidine tag at the C-terminus. Proline substitutions (F817P/ A892P/ A899P/ A942P/ K986P/ V987P) and alanine substitutions (R683A and R685A) are introduced to stabilize the trimeric prefusion state of SARS-CoV-2 S protein and abolish the furin cleavage site, respectively. HV69-70del/ Y144del/ N501Y/ A570D/ D614G/ P681H/ T716I/ S982A/ D1118H/ mutations were identified on the spike protein in the SARS-CoV-2 variant (known as B.1.1.7, 20B/501Y.V1 or VOC 202012/01) which emerged in the United Kingdom.;This protein carries a polyhistidine tag at the C-terminus, followed by an Avi tag. The protein has a calculated MW of 139.4 kDa. The protein migrates as 160-200 kDa under reducing (R) con
Catalog No. 50-210-7327 Supplier ACROBiosystems Supplier No. SPNC82E5200UG
Recombinant Protein;200UG;Biotinylated SARS-CoV-2 S protein trimer, His,Avitag (SPN-C82E5) is expressed from human 293 cells (HEK293). It contains AA Val 16 - Pro 1213 (Accession # QHD43416.1). The recombinant protein is expressed from human 293 cells (HEK293) with T4 fibritin trimerization motif and a polyhistidine tag at the C-terminus. Proline substitutions (F817P/ A892P/ A899P/ A942P/ K986P/ V987P) and alanine substitutions (R683A and R685A) are introduced to stabilize the trimeric prefusion state of SARS-CoV-2 S protein and abolish the furin cleavage site, respectively. HV69-70del/ Y144del/ N501Y/ A570D/ D614G/ P681H/ T716I/ S982A/ D1118H/ mutations were identified on the spike protein in the SARS-CoV-2 variant (known as B.1.1.7, 20B/501Y.V1 or VOC 202012/01) which emerged in the United Kingdom.;This protein carries a polyhistidine tag at the C-terminus, followed by an Avi tag. The protein has a calculated MW of 139.4 kDa. The protein migrates as 160-200 kDa under reducing (R) con