Learn More
ACROBiosystems Biotinylated SARS-CoV-2 S protein (D614G), His,Avitag™, Super stable trimer (MALS verified)
Supplier:  ACROBiosystems SPNC82E3200UG
Encompass_Preferred
Recombinant Protein;200UG;Biotinylated SARS-CoV-2 S protein (D614G), His,Avitag, Super stable trimer (SPN-C82E3) is the ectodomain of SARS-CoV-2 S protein that contains AA Val 16 - Pro 1213 (Accession # QHD43416.1 (D614G)) and D614G mutation, which has become increasingly common in SARS-CoV-2 viruses from around the world . The recombinant protein is expressed from human 293 cells (HEK293) with T4 fibritin trimerization motif and a polyhistidine tag at the C-terminus. Proline substitutions (F817P, A892P, A899P, A942P, K986P, V987P) and alanine substitutions (R683A and R685A) are introduced to stabilize the trimeric prefusion state of SARS-CoV-2 S protein and abolish the furin cleavage site, respectively. It is the biotinlynated form of SARS-CoV-2 S protein (D614G), His Tag, Super stable trimer (MALS verified) (Cat. No. SPN-C52H3).;This protein carries a polyhistidine tag at the C-terminus, followed by an Avi tag. The protein has a calculated MW of 139.7 kDa. The protein migrates as 15
Catalog No. 50-205-5146