Learn More
ACROBiosystems ACROBiosystems Biotinylated SARS-CoV-2 Spike RBD (L452R, E484Q), His,Avitag™ (MALS verified)
Catalog No. 502107040
Encompass_Preferred
Recombinant Protein;200UG;Biotinylated SARS-CoV-2 Spike RBD (L452R, E484Q), His,Avitag (SPD-C82Ec) is expressed from human 293 cells (HEK293). It contains AA Arg 319 - Lys 537 (Accession # QHD43416.1 (L452R, E484Q)). The L452R, E484Q mutations were identified in the SARS-CoV-2 variants which emerged in India (known as B.1.617). ;This protein carries a polyhistidine tag at the C-terminus, followed by an Avi tag. The protein has a calculated MW of 28.3 kDa. The protein migrates as 33-45 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Human ACE2, Fc Tag (Cat. No. AC2-H5257) at 2 μg/mL (100 μL/well) can bind Biotinylated SARS-CoV-2 Spike RBD (L452R, E484Q), His,Avitag (Cat. No. SPD-C82Ec) with a linear range of 10-78 ng/mL (QC tested).;Immobilized Biotinylated SARS-CoV-2 Spike RBD (L452R, E484Q), His,Avitag (Cat. No. SPD-C82Ec) at 1 μg/mL (100 μL/well) on streptavidin (Cat. No. STN-N5116) precoated (0.5 μg/well) plate can bind Human ACE2, Fc Tag (Cat. No. AC2-
Catalog No. 50-210-7040 Supplier ACROBiosystems Supplier No. SPDC82EC200UG
Recombinant Protein;200UG;Biotinylated SARS-CoV-2 Spike RBD (L452R, E484Q), His,Avitag (SPD-C82Ec) is expressed from human 293 cells (HEK293). It contains AA Arg 319 - Lys 537 (Accession # QHD43416.1 (L452R, E484Q)). The L452R, E484Q mutations were identified in the SARS-CoV-2 variants which emerged in India (known as B.1.617). ;This protein carries a polyhistidine tag at the C-terminus, followed by an Avi tag. The protein has a calculated MW of 28.3 kDa. The protein migrates as 33-45 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Human ACE2, Fc Tag (Cat. No. AC2-H5257) at 2 μg/mL (100 μL/well) can bind Biotinylated SARS-CoV-2 Spike RBD (L452R, E484Q), His,Avitag (Cat. No. SPD-C82Ec) with a linear range of 10-78 ng/mL (QC tested).;Immobilized Biotinylated SARS-CoV-2 Spike RBD (L452R, E484Q), His,Avitag (Cat. No. SPD-C82Ec) at 1 μg/mL (100 μL/well) on streptavidin (Cat. No. STN-N5116) precoated (0.5 μg/well) plate can bind Human ACE2, Fc Tag (Cat. No. AC2-