Learn More
Vector Laboratories Lycopersicon esculentum lectin (LEL, TL) Biotinylated 1 mg
SDP
Catalog No. NC9862633
Encompass
Lycopersicon esculentum lectin (LEL or tomato lectin, TL) binds well to glycophorin and Tamm-Horsfall glycoprotein and has been used effectively to label vascular endothelium in rodents. This conjugate features a ratio of biotin to lectin protein that provides optimal avidin/streptavidin and lectin binding activity. Supplied essentially free of unconjugated biotins, it is preserved with sodium azide.
Catalog No. NC9862633 Supplier Vector Laboratories Supplier No. B11751
Lycopersicon esculentum lectin (LEL or tomato lectin, TL) binds well to glycophorin and Tamm-Horsfall glycoprotein and has been used effectively to label vascular endothelium in rodents. This conjugate features a ratio of biotin to lectin protein that provides optimal avidin/streptavidin and lectin binding activity. Supplied essentially free of unconjugated biotins, it is preserved with sodium azide.