Learn More
Vector Laboratories Wisteria floribunda lectin (WFA, WFL) Biotinylated 2 mg
SDP
Supplier:  Vector Laboratories B13552
Encompass
Though the binding specificity of Wisteria floribunda lectin (WFL) is not entirely clear, this lectin appears to preferentially bind carbohydrate structures terminating in N-acetylgalactosamine linked α or β to the 3 or 6 position of galactose. WFL has been used to fractionate lymphocyte populations, and although it is not mitogenic, it elicits the production of lymphokines from murine splenocytes. This conjugate features a ratio of biotin to lectin protein that provides optimal avidin/streptavidin and lectin binding activity. Supplied essentially free of unconjugated biotins, it is preserved with sodium azide.
Catalog No. NC1042310