Learn More
ACROBiosystems ACROBiosystems Biotinylated Zika virus NS1 Protein, Avitag™,His Tag
Catalog No. 502106730
Encompass_Preferred
Recombinant Protein;25UG;Biotinylated Zika virus NS1, Avitag,His Tag (NS1-Z82E9) is expressed from human 293 cells (HEK293). It contains AA Val 796 - Ser 1148 (Accession # ALU33341.1).;This protein carries an Avi tag (Avitag™) at the C-terminus, followed by a polyhistidine tag. The protein has a calculated MW of 43.3 kDa. The protein migrates as 46-50 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.
Catalog No. 50-210-6730 Supplier ACROBiosystems Supplier No. NS1Z82E925UG
Recombinant Protein;25UG;Biotinylated Zika virus NS1, Avitag,His Tag (NS1-Z82E9) is expressed from human 293 cells (HEK293). It contains AA Val 796 - Ser 1148 (Accession # ALU33341.1).;This protein carries an Avi tag (Avitag™) at the C-terminus, followed by a polyhistidine tag. The protein has a calculated MW of 43.3 kDa. The protein migrates as 46-50 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.