Learn More
Abcam Biotinylation Kit / Biotin Conjugation Kit (Fast, Type A) - Lightning-Link, 1 mg, 1 reaction, optimized for IgG antibodies, uses primary amine groups
Catalog No. NC2077085
Encompass_Preferred
Biotinylation Kit / Biotin Conjugation Kit (Fast, Type A) - Lightning-Link, 1 mg, 1 reaction, optimized for IgG antibodies, uses primary amine groups
Catalog No. NC2077085 Supplier Abcam Supplier No. AB2017951MG
Biotinylation Kit / Biotin Conjugation Kit (Fast, Type A) - Lightning-Link, 1 mg, 1 reaction, optimized for IgG antibodies, uses primary amine groups