Learn More
Abcam Biotinylation Kit / Biotin Conjugation Kit (Fast, Type B) - Lightning-Link, 3 x 100UG
Catalog No. 502589155
Encompass_Preferred
Biotin conjugation / labeling in < 20 mins with 30 secs hands-on time using Biotinylation Kit / Biotin Conjugation Kit (Fast, Type B) - Lightning-Link ab201796. 100% antibody recovery - Cited in over 15 publications - Biotin labeled antibody is ready for use in applications like WB, ELISA, and IHC, with no need for further purification - Compatible with most standard antibody formulations - Scalable;use the same conjugation method from 10µg-100mg of antibody - Use Lightning-Link conjugation with over 45 labels, including Alexa Fluor, DyLight, tandem dyes, enzymes, oligonucleotides, and gold nanoparticles
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-258-9155 Supplier Abcam Supplier No. AB2017963X100UG
Biotin conjugation / labeling in < 20 mins with 30 secs hands-on time using Biotinylation Kit / Biotin Conjugation Kit (Fast, Type B) - Lightning-Link ab201796. 100% antibody recovery - Cited in over 15 publications - Biotin labeled antibody is ready for use in applications like WB, ELISA, and IHC, with no need for further purification - Compatible with most standard antibody formulations - Scalable;use the same conjugation method from 10µg-100mg of antibody - Use Lightning-Link conjugation with over 45 labels, including Alexa Fluor, DyLight, tandem dyes, enzymes, oligonucleotides, and gold nanoparticles
The product is subject to the following: Abcam Restricted Use Statement