Learn More
Abcam Biotinylation Kit / Biotin Conjugation Kit (Fast, Type B) - Lightning-Link, 1 mg, reaction time <20 min, hands-on time 30 sec, 100% antibody recovery, no purification needed
Catalog No. NC2326296
Encompass_Preferred
The Abcam Biotinylation Kit provides a rapid and efficient method for biotinylating antibodies and proteins, allowing for fast labeling in laboratory settings. With minimal hands-on time, this kit ensures total antibody recovery without the need for purification, making it ideal for various downstream applications including Western Blot, ELISA, and Immunohistochemistry.
  • Biotin labeling in under 20 minutes
  • Minimal hands-on time of 30 seconds
  • 100% antibody recovery
  • Targets primary amine groups like lysines
  • Recommended antibody amount of 1 mg
  • Maximum antibody amount of 2 mg
  • Suitable for buffer pH of 6.5 to 8.5
  • No purification required
  • Compatible with Western Blot, ELISA, and IHC applications
  • Ideal antibody concentration of 1 mg/mL
  • Kit size of 1 mg
  • Storage temperature of -20°C
Catalog No. NC2326296 Supplier Abcam Supplier No. AB2017961MG
The Abcam Biotinylation Kit provides a rapid and efficient method for biotinylating antibodies and proteins, allowing for fast labeling in laboratory settings. With minimal hands-on time, this kit ensures total antibody recovery without the need for purification, making it ideal for various downstream applications including Western Blot, ELISA, and Immunohistochemistry.
  • Biotin labeling in under 20 minutes
  • Minimal hands-on time of 30 seconds
  • 100% antibody recovery
  • Targets primary amine groups like lysines
  • Recommended antibody amount of 1 mg
  • Maximum antibody amount of 2 mg
  • Suitable for buffer pH of 6.5 to 8.5
  • No purification required
  • Compatible with Western Blot, ELISA, and IHC applications
  • Ideal antibody concentration of 1 mg/mL
  • Kit size of 1 mg
  • Storage temperature of -20°C