Learn More
ACROBiosystems Biotinylated Mouse Monoclonal Antibody, 200 μg, Targets DXD and Exatecan, Mouse IgG1, Kappa Isotype, Lyophilized, Compatible with ELISA
Catalog No. NC3370907
Encompass_Preferred
This technical summary describes a biotinylated mouse monoclonal antibody developed by ACROBiosystems, specifically for detecting DXD and Exatecan. The antibody is expressed in HEK293 cells and provides high specificity and consistent performance in ELISA applications. Its biotinylation permits versatile detection using streptavidin-conjugated reagents, making it an invaluable tool for research involving Exatecan ADC derivatives. The product is lyophilized for long-term stability when stored correctly.
  • Detects DXD target
  • Recombinantly expressed in HEK293 cells
  • Protein A/G purified
  • Lyophilized format ensures stability
  • Recommended application: ELISA
  • Recommended ELISA concentration: 1-5000 ng/mL
  • Long-term storage: -20°C to -70°C for 12 months
  • Quantity: 200 μg
Catalog No. NC3370907 Supplier ACROBiosystems Supplier No. DXDBVM807200UG
This technical summary describes a biotinylated mouse monoclonal antibody developed by ACROBiosystems, specifically for detecting DXD and Exatecan. The antibody is expressed in HEK293 cells and provides high specificity and consistent performance in ELISA applications. Its biotinylation permits versatile detection using streptavidin-conjugated reagents, making it an invaluable tool for research involving Exatecan ADC derivatives. The product is lyophilized for long-term stability when stored correctly.
  • Detects DXD target
  • Recombinantly expressed in HEK293 cells
  • Protein A/G purified
  • Lyophilized format ensures stability
  • Recommended application: ELISA
  • Recommended ELISA concentration: 1-5000 ng/mL
  • Long-term storage: -20°C to -70°C for 12 months
  • Quantity: 200 μg