Learn More
ACROBiosystems BIOTINYLTD HM PD-L1/CD274,25UG
Supplier: ACROBiosystems PD1H82F325UG
MABSol Biotinylated Human PD-L1 / B7-H1, His Tag & Fc Tag (PD1-H82F3) is expressed from human HEK293 cells. It contains AA Phe 19 - Arg 238 (Accession # NP_054862.1).