Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bio Plas™ BioTip™ Low Retention Pipette Tips, Bulk

Catalog No. 21402694
Change view
Click to view available options
Volume (Metric):
10 μL
200 μL
1000 μL
1250 μL
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Volume (Metric)
21-402-694 1250 μL
21-402-680 10 μL
21-402-686 200 μL
21-402-691 1000 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 21-402-694 Supplier Bio Plas™ Supplier No. 3105B
Add to Cart
Add to Cart

Bulk supply, natural color, polypropylene construction, made in the USA

  • Low retention surface minimizes sample loss and improves accuracy, especially for small volumes and critical assays
  • Bulk packaging format allows for efficient storage and cost-effective high-volume laboratory use
  • Natural color design enables easy visibility of liquids during pipetting procedures
  • Constructed from high-quality polypropylene for durability and chemical resistance
  • Manufactured in the USA, ensuring consistent quality and reliable performance

Specifications

Volume (Metric) 1250 μL
Filtered Non-filtered
Sterility Non-sterile
Color Clear
Autoclavable Autoclavable
Format Bulk
Tip Style Flat
Quantity 1000 Tips/Bag
Product Type Pipette Tip
Color Code White
Disposable Single-use
For Use With (Equipment) Universal Fit
Graduated Yes
Material 100% Virgin Polypropylene
Fit for Purpose Certified Quality
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.