Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bio Plas™ BioTip™ Low Retention Pipette Tips, EnviroPak Refills

Catalog No. 21402681
Change view
Click to view available options
Volume (Metric):
10 μL
200 μL
1250 μL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Volume (Metric)
21-402-681 10 μL
21-402-687 200 μL
21-402-695 1250 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 21-402-681 Supplier Bio Plas™ Supplier No. 3100E
Only null left
Add to Cart
Add to Cart

Low retention pipette tips with EnviroPak refill system

  • EnviroPak refill system supports environmentally conscious lab operations by minimizing packaging waste
  • Low retention design enhances sample recovery and improves pipetting accuracy
  • Racked format with 96 tips per rack allows for convenient handling and organized workflow
  • Constructed from durable natural polypropylene for reliable chemical resistance and performance
  • Manufactured in the USA to ensure consistent quality standards

Specifications

Volume (Metric) 10 μL
Filtered Non-filtered
Sterility Non-sterile
Color Clear
Autoclavable Autoclavable
Format Refill
Tip Style Flat
Product Type Pipette Tip
Color Code White
Disposable Single-use
For Use With (Equipment) Universal Fit
Graduated Yes
Material 100% Virgin Polypropylene
Fit for Purpose Certified Quality
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.